Learn More
Sino Biological Recombinant Human B3GALTL Protein (His Tag)
Supplier:  Sino Biological 13957H07H100
Encompass_Preferred
A DNA sequence encoding the human B3GALTL (JC8008) (Ser29-Phe494) was expressed with an N-terminal polyhistidine tag.
Catalog No. 50-161-8961