Learn More
eMolecules​ 139631-62-2 | Cyclopropanesulphonyl chloride | Apollo Scientific | MFCD01631933 | 140.580 | C3H5ClO2S | 95.000 | ClS(=O)(=O)C1CC1 | 25g | 562447063
Catalog No. 5000490656 Shop All eMolecules Inc. Products
Encompass_Preferred
Cyclopropanesulphonyl chloride | Apollo Scientific | 139631-62-2 | MFCD01631933 | 140.580 | C3H5ClO2S | 95.000 | ClS(=O)(=O)C1CC1 | 25g | 562447063
Catalog No. 50-004-90656 Supplier eMolecules​ Supplier No. 562447063
Cyclopropanesulphonyl chloride | Apollo Scientific | 139631-62-2 | MFCD01631933 | 140.580 | C3H5ClO2S | 95.000 | ClS(=O)(=O)C1CC1 | 25g | 562447063