Learn More
Sino Biological Recombinant Human ROR1 Protein (hFc Tag)
Supplier:  Sino Biological 13968H02H1100
Encompass_Preferred
A DNA sequence encoding the human ROR1 (NP_005003.2)(Met1-Glu403) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8963