Learn More
Sino Biological Recombinant Human ROR1 Protein (aa 453-783, His & GST Tag)
Supplier:  Sino Biological 13968H20B50
Encompass_Preferred
A DNA sequence encoding the human ROR1 (AAA60275.1) (Met453-Asn783) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-8968