Learn More
Sino Biological Recombinant Human PLAC9 Protein (mFc Tag)
Supplier:  Sino Biological 13971H05H100
Encompass_Preferred
A DNA sequence encoding the huamn PLAC9 (Q5JTB6) (Met1-Phe97) was fused with Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-8972