Learn More
Cayman Chemical 13ACtyl9DIhydrobaccatIn I 10mg
SDP
Catalog No. 502989127
Encompass_Preferred
A taxane with anticancer activity; inhibits proliferation of P388 leukemia cells (IC50 = 20 μg/ml)
Catalog No. 50-298-9127 Supplier Cayman Chemical Supplier No. 2970110 MG
A taxane with anticancer activity; inhibits proliferation of P388 leukemia cells (IC50 = 20 μg/ml)