Learn More
Cayman Chemical 13C17-Mycophenolic Acid, Purity: ≥95%, CAS: 1202866-92-9, 1.2 mL
SDP
Catalog No. 502912153
Encompass_Preferred
An internal standard for the quantification of mycophenolic acid by GC- or LC-MS
Catalog No. 50-291-2153 Supplier Cayman Chemical Supplier No. 363591.2 ML
An internal standard for the quantification of mycophenolic acid by GC- or LC-MS