Learn More
Carbosynth LLC. [13C18,15N3]-Hepcidin (Human) (0.02mg vial), 1ea, Disulfide bonds: Reported disulfide bonds between Cys7-Cys23, Cys10-Cys13, Cys11-Cys19, and Cys14-Cys22, MW: 2810.2, Molecular formula: C9513C18H170N3115N3O31S9
SDP
Supplier:  Carbosynth LLC. PLP3405V0.02MG
Encompass_Preferred
Synonyms: Asp-Thr-His-[13C9,15N]Phe-Pro-Ile-Cys-Ile-[13C9,15N]Phe-Cys-Cys-[15N]Gly-Cys-Cys-His-Arg-Ser-Lys-Cys-Gly-Met-Cys-Cys-Lys-Thr; [[13C9,15N]Phe4,9, [15N]Gly12]-Hepcidin (Human), Application: Stable Isotope-Labeled Peptide for Mass Spectrometric Detection of Hepcidin (Human), Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20°C, References: N. Murao, M. Ishigai, H. Yasuno, Y. Shimonaka, and Y. Aso, Rapid Commun. Mass Spectrom., 21, 4033 (2007)., T. Hosoki, K. Ikuta, Y. Shimonaka, Y. Sasaki, H. Yasuno, K. Sato, T. Ohtake, K. Sasaki, Y. Torimoto, K. Saito, and Y. Kohgo, Proteomics Clin. Appl., 3, 1256 (2009).
Catalog No. 50-168-6593