Learn More
Cayman Chemical 13C28-Bongkrekic Acid, Purity: ≥98%, 1.2 mL
SDP
Catalog No. 502915659
Encompass_Preferred
An internal standard for the quantification of bongkrekic acid by GC- or LC-MS
Catalog No. 50-291-5659 Supplier Cayman Chemical Supplier No. 394541.2 ML
An internal standard for the quantification of bongkrekic acid by GC- or LC-MS