Learn More
Cayman Chemical 13C6-4-Nitroaniline, Purity: ≥98%, CAS: 1028361-70-7, 10 mg
SDP
Catalog No. 502910914
Encompass_Preferred
An internal standard for the quantification of 4-nitroaniline by GC- or LC-MS
Catalog No. 50-291-0914 Supplier Cayman Chemical Supplier No. 3485810 MG
An internal standard for the quantification of 4-nitroaniline by GC- or LC-MS