Learn More
Cayman Chemical 13C6-3-Nitrophenylhydrazine (hydrochloride), Purity: ≥98%, CAS: 1977535-33-3, 10 mg
SDP
Catalog No. 502984255
Encompass_Preferred
A 13C-labeled derivatizing reagent for carboxylic acids
Catalog No. 50-298-4255 Supplier Cayman Chemical Supplier No. 2074410 MG
A 13C-labeled derivatizing reagent for carboxylic acids