Learn More
Cayman Chemical 13C6-4-Nitrophenylhydrazine (hydrochloride), Purity: ≥95%, 10 mg
SDP
Catalog No. 502945777
Encompass_Preferred
An internal standard for the quantification of 4-nitrophenylhydrazine by GC- or LC-MS; a byproduct in the synthesis of 13C6-3-nitrophenylhydrazine
Catalog No. 50-294-5777 Supplier Cayman Chemical Supplier No. 4047910 MG
An internal standard for the quantification of 4-nitrophenylhydrazine by GC- or LC-MS; a byproduct in the synthesis of 13C6-3-nitrophenylhydrazine