Learn More
Cayman Chemical 13epi12oxo Phytodienoic Acid
SDP
Catalog No. 502887179
Encompass_Preferred
13-epi-12-oxo PDA is an LO metabolite of α-linolenic acid in the leaves of green plants such as corn. ω-3 and ω-6 PUFAs in plants are substrates for plant LOs.{857} 12-oxo PDA is one of the best studied end metabolites of this enzymatic pathway.{1562} While the initial enzymatic product and major isomer of 12-oxo PDA contains side chains in the cis position, both being β to the ring, the upper side chain attached at C-13, can and frequently does, isomerize when 12-oxo PDA is extracted, isolated, or stored. 13-epi-12-oxo PDA is the product of this isomerization.
Catalog No. 50-288-7179 Supplier Cayman Chemical Supplier No. 10195100 UG
Add to Cart
Add to Cart
13-epi-12-oxo PDA is an LO metabolite of α-linolenic acid in the leaves of green plants such as corn. ω-3 and ω-6 PUFAs in plants are substrates for plant LOs.{857} 12-oxo PDA is one of the best studied end metabolites of this enzymatic pathway.{1562} While the initial enzymatic product and major isomer of 12-oxo PDA contains side chains in the cis position, both being β to the ring, the upper side chain attached at C-13, can and frequently does, isomerize when 12-oxo PDA is extracted, isolated, or stored. 13-epi-12-oxo PDA is the product of this isomerization.