Learn More
Essendant Scrubbing Floor Pads, 13"" Diameter, Blue, 5/Carton
Catalog No. 502003381
Encompass_Preferred
For heavy-duty scrubbing prior to recoating; Removes dirt, spills and scuffs, leaving a clean surface ready for recoating. Use with rotary or automatic machines (Speed of 175-600 rpms). Color-coded for easy job identification.
Catalog No. 50-200-3381 Supplier Essendant Supplier No. BWK4013BLU
For heavy-duty scrubbing prior to recoating; Removes dirt, spills and scuffs, leaving a clean surface ready for recoating. Use with rotary or automatic machines (Speed of 175-600 rpms). Color-coded for easy job identification.