Learn More
Chemglass Life Sciences 13" One-Piece PTFE Lower Agitator, Four Blade, For Use with 30mm Shafts, For use with 400mm ID Flange Reactor
SDP
Catalog No. 501531445
Encompass_Preferred
13" One-Piece PTFE Lower Agitator, Four Blade, For Use with 30mm Shafts, For use with 400mm ID Flange Reactor. High viscosity agitator. The body is made entirely from ONE PIECE of CLASS VI VIRGIN PTFE. One-piece machined body is much easier to clean. The smooth machined surface has no seams, joints or crevices for material to collect. The design provides excellent stirring at the bottom of CG-1968-W 400mm ID 100L reactor vessels, especially when heavy slurries are being mixed. Agitator attaches to stir shaft by both a compression fitting and a PTFE safety pin. Heavy duty blades 9mm thick. For use with 30mm CG-2097 stirrer shafts. PLEASE NOTE: THE LOWER AGITATOR HAS BUSHING TO ACCEPT A STABILIZING PIN FROM DRAIN VALVE PLUG. THE PIN EXTENDS INTO THE LOWER AGITATOR AND VIRTUALLY ELIMINATES ANY WHIPPING BY STABILIZING THE STIRRER SHAFT.
Catalog No. 50-153-1445 Supplier Chemglass Life Sciences Supplier No. CG2095M55
13" One-Piece PTFE Lower Agitator, Four Blade, For Use with 30mm Shafts, For use with 400mm ID Flange Reactor. High viscosity agitator. The body is made entirely from ONE PIECE of CLASS VI VIRGIN PTFE. One-piece machined body is much easier to clean. The smooth machined surface has no seams, joints or crevices for material to collect. The design provides excellent stirring at the bottom of CG-1968-W 400mm ID 100L reactor vessels, especially when heavy slurries are being mixed. Agitator attaches to stir shaft by both a compression fitting and a PTFE safety pin. Heavy duty blades 9mm thick. For use with 30mm CG-2097 stirrer shafts. PLEASE NOTE: THE LOWER AGITATOR HAS BUSHING TO ACCEPT A STABILIZING PIN FROM DRAIN VALVE PLUG. THE PIN EXTENDS INTO THE LOWER AGITATOR AND VIRTUALLY ELIMINATES ANY WHIPPING BY STABILIZING THE STIRRER SHAFT.