Learn More
Cayman Chemical 13methyl PentadecnoIc ACD 10mg
SDP
Catalog No. 502932567
Encompass_Preferred
A methylated fatty acid methyl ester; has been found in the bioactive fraction of A. malabarica extract, C. variabilis microalgae, and baboon liver
Catalog No. 50-293-2567 Supplier Cayman Chemical Supplier No. 2481910 MG
A methylated fatty acid methyl ester; has been found in the bioactive fraction of A. malabarica extract, C. variabilis microalgae, and baboon liver