Learn More
Cayman Chemical 13-methyl Pentadecanoic Acid methyl ester, Purity: ≥98%, CAS: 5487-50-3, 20 mg
SDP
Catalog No. 502932568
Encompass_Preferred
A methylated fatty acid methyl ester; has been found in the bioactive fraction of A. malabarica extract, C. variabilis microalgae, and baboon liver
Catalog No. 50-293-2568 Supplier Cayman Chemical Supplier No. 2481920 MG
A methylated fatty acid methyl ester; has been found in the bioactive fraction of A. malabarica extract, C. variabilis microalgae, and baboon liver