Learn More
Sarstedt Inc 13ML SCTUBE 16.5X101 RBS/CS500
Supplier:  Sarstedt Inc 60.540.386
Encompass_Preferred
13mL Screw Cap Tube 16.5x101mm Flat Base Polypropylene Neutral Cap Assembled Sterile
Catalog No. NC9054766