Learn More
Sarstedt Inc 13ML SCTUBE 16.5X101RBS/PK1000
Catalog No. NC9437081 Shop All Sarstedt Inc. Products
Encompass_Preferred
13mL Screw Cap Tube 16.5x101mm Round Bottom Skirted Polypropylene Neutral Cap
Catalog No. NC9437081 Supplier Sarstedt Inc Supplier No. 60.541.385
13mL Screw Cap Tube 16.5x101mm Round Bottom Skirted Polypropylene Neutral Cap