Learn More
Sarstedt Inc 13ML TUBE 16.5X101ORING/PK1000
Catalog No. NC9080781 Shop All Sarstedt Inc. Products
Encompass_Preferred
Tubes, 13mL, 16.5x101mm, Round Bottom, Polypropylene, O-Ring Screw Cap
Catalog No. NC9080781 Supplier Sarstedt Inc Supplier No. 60.541.304
Tubes, 13mL, 16.5x101mm, Round Bottom, Polypropylene, O-Ring Screw Cap