Learn More
Analytical Sales & Services 13mm, 15 x 45mm, 3.1mL Clear Glass Screw High Recovery MicroVials
Catalog No. 5000807014
Encompass
Advantage™ 13mm, 15 x 45mm, 3.1mL Volume Clear Glass Screw Thread High Recovery MicroVials - for use with Shimadzu and Waters WISP 48 Position Autosamplers
Catalog No. 50-008-07014 Supplier Analytical Sales & Services Supplier No. 31561
Add to Cart
Edge
Add to Cart
Advantage™ 13mm, 15 x 45mm, 3.1mL Volume Clear Glass Screw Thread High Recovery MicroVials - for use with Shimadzu and Waters WISP 48 Position Autosamplers