Learn More
Analytical Sales & Services 13mm, 15 x 45mm, 4mL Amber Glass Screw Vials
Catalog No. 5000804462
Encompass
Advantage™ 13mm, 15 x 45mm, 4mL Amber Glass Screw Vials - for Shimadzu and Waters WISP 48 Position Autosamplers
Catalog No. 50-008-04462 Supplier Analytical Sales & Services Supplier No. 31532CASE
Advantage™ 13mm, 15 x 45mm, 4mL Amber Glass Screw Vials - for Shimadzu and Waters WISP 48 Position Autosamplers