Learn More
Analytical Sales & Services 13mm, 15 x 45mm, 4mL Clear Glass Screw Vials with Marking Spots
Catalog No. 5000806059
Encompass
Advantage™ 13mm, 15 x 45mm, 4mL Clear Glass Screw Vials with Marking Spots - for Shimadzu and Waters WISP 48 Position Autosamplers
Catalog No. 50-008-06059 Supplier Analytical Sales & Services Supplier No. 31531MCASE
Advantage™ 13mm, 15 x 45mm, 4mL Clear Glass Screw Vials with Marking Spots - for Shimadzu and Waters WISP 48 Position Autosamplers