Learn More
Analytical Sales & Services 13mm, 15 x 45mm, 4mL Clear Glass Snap and Seal Vials
Catalog No. 5000806874
Encompass
Advantage™ 13mm, 15 x 45mm, 4mL Clear Glass Snap and Seal Vials - for Shimadzu Autosamplers
Catalog No. 50-008-06874 Supplier Analytical Sales & Services Supplier No. 31511CASE
Advantage™ 13mm, 15 x 45mm, 4mL Clear Glass Snap and Seal Vials - for Shimadzu Autosamplers