Learn More
Analytical Sales & Services 13mm, 4mL Amber Screw Vials and Black PP Caps with 0.60" Silicone/PTFE Liners
Catalog No. 5000804838
Encompass
Advantage™ Unassembled Vial Kit. Contains: 13mm, 4mL Amber Glass Screw Vials and Black Polypropylene Caps with 0.60" Silicone/PTFE Liners 1000/CASE
Catalog No. 50-008-04838 Supplier Analytical Sales & Services Supplier No. 53631ACASE
Advantage™ Unassembled Vial Kit. Contains: 13mm, 4mL Amber Glass Screw Vials and Black Polypropylene Caps with 0.60" Silicone/PTFE Liners 1000/CASE