Learn More
Analytical Sales & Services 13mm, 4mL Amber Screw Vials and Black PP Caps with Pre-Slit Silicone/PTFE Liners
Catalog No. 5000804316
Encompass
Advantage™ Assembled Vial Kit\n\nContains:\n• 13mm, 4mL Amber Glass Screw Vials\n• Black Polypropylene Caps with Pre-Slit 0.65" Red Silicone/PTFE Liners\n\nQty/Pkg: 1000
Catalog No. 50-008-04316 Supplier Analytical Sales & Services Supplier No. 51526SLCASE
Advantage™ Assembled Vial Kit\n\nContains:\n• 13mm, 4mL Amber Glass Screw Vials\n• Black Polypropylene Caps with Pre-Slit 0.65" Red Silicone/PTFE Liners\n\nQty/Pkg: 1000