Learn More
Analytical Sales & Services 13mm, 4mL Clear Screw Vials and Black PP Caps with 0.65" Red Silicone/PTFE Liners
Catalog No. 5000805694
Encompass
Advantage™ Assembled Vial Kit. Contains: 13mm, 4mL Clear Glass Screw Vials and Black Polypropylene Caps with 0.65" Red Silicone/PTFE Liners
Catalog No. 50-008-05694 Supplier Analytical Sales & Services Supplier No. 51516CASE
Advantage™ Assembled Vial Kit. Contains: 13mm, 4mL Clear Glass Screw Vials and Black Polypropylene Caps with 0.65" Red Silicone/PTFE Liners