Learn More
Analytical Sales & Services 13mm, 4mL Clear Screw Vials and Black PP Caps with 0.75" Silicone/PTFE Liners
Catalog No. 5000807073
Encompass
Advantage™ Unassembled Vial Kit. Contains: 13mm, 4mL Clear Glass Screw Vials and Black Polypropylene Caps with 0.75" Silicone/PTFE Liners
Catalog No. 50-008-07073 Supplier Analytical Sales & Services Supplier No. 53731CASE
Add to Cart
Add to Cart
Advantage™ Unassembled Vial Kit. Contains: 13mm, 4mL Clear Glass Screw Vials and Black Polypropylene Caps with 0.75" Silicone/PTFE Liners