Learn More
Analytical Sales & Services 13mm, 4mL Clear Screw Vials and Black PP Caps with Pre-Slit 0.65" Red Silicone/PTFE Liners
Catalog No. 5000804978
Encompass
Advantage™ Assembled Vial Kit. Contains: 13mm, 4mL Clear Glass Screw Vials and Black Polypropylene Caps with Pre-Slit 0.65" Red Silicone/PTFE Liners
Catalog No. 50-008-04978 Supplier Analytical Sales & Services Supplier No. 51516SLCASE
Advantage™ Assembled Vial Kit. Contains: 13mm, 4mL Clear Glass Screw Vials and Black Polypropylene Caps with Pre-Slit 0.65" Red Silicone/PTFE Liners