Learn More
Cytiva Acrodisc Syringe Filter with wwPTFE Membrane - 0.45 µm, 13 mm, minispike outlet (100/pkg 300/cs)
Supplier:  Cytiva 2402
Encompass
Syringe filters with wwPTFE membrane are ideal for the filtration of a wide variety of common solvents including aqueous and organic solvents used in HPLC (0.45 μm) and UHPLC (0.2 μm) sample prep. Filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending chromatography column life and reducing system downtime. The syringe filters are low in protein and API binding which make them ideal for drug studies, such as dissolution testing. Both the membrane and housing are extremely low in levels of potential extractables.13 mm Acrodisc syringe filters are designed for the filtration of sample volumes up to 10 mL. The syringe filters are HPLC certified for low levels of UV-absorbing extractables, ensuring accurate and reliable results.Minispike outlet offer easy handling with a HPLC vial and reduced sample hold-up volume. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1749180