Learn More
Cytiva (Formerly Pall Lab) Acrodisc Syringe Filter with PTFE Membrane - 0.45 m, 13 mm, minispike outlet (100/pkg 300/cs)
Supplier:  Cytiva 4422
Encompass
Acrodisc Syringe Filters with PTFE Membrane - 0.45 m, 13mm, male slip luer outlet.Syringe filters with PTFE membrane are ideal for the filtration of organic solvents and harsh solvents in HPLC (0.45 m) and UHPLC (0.2 m) sample preparation. Sample filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending the life of chromatography columns and reducing system downtime. PTFE is a hydrophobic membrane and is not suitable for the filtration of aqueous solutions, however these syringe filters are ideal to use in venting applications. 13 mm Acrodisc syringe filters are designed for the filtration of sample volumes up to 10 mL. The syringe filters are HPLC certified for low levels of UV-absorbing extractables, ensuring accurate and reliable results. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1699843