Learn More
Across International 13mm Diameter ID Dry Pressing Die Set
SDP
Catalog No. 502251331
Encompass_Preferred
A set of 13mm diameter (ID) hardened steel dry pressing die set for pellet presses, ready for you to make 13mm diameter cylinder-shape pellets
This item is not returnable. View return policy
Catalog No. 50-225-1331 Supplier Across International Supplier No. SDS13
A set of 13mm diameter (ID) hardened steel dry pressing die set for pellet presses, ready for you to make 13mm diameter cylinder-shape pellets