Learn More
Ace Glass, Inc. 13MM GRADED SEALS 13MM GRADED SEALS
SDP
Catalog No. 50974597
Encompass
13mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-597 Supplier Ace Glass, Inc. Supplier No. 845522
13mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.