Learn More
Glass Expansion 13mm Metal Ball Joint Clip
SDP
Catalog No. 502702975
Encompass_Preferred
13mm Metal Ball Joint Clip. Used to secure 13mm male and female ball joints for torches, transfer tubes and spray chambers. For 19mm ball joints use JC-19.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2975 Supplier Glass Expansion Supplier No. JC13
13mm Metal Ball Joint Clip. Used to secure 13mm male and female ball joints for torches, transfer tubes and spray chambers. For 19mm ball joints use JC-19.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001