Learn More
Labcon North America TipTop Dispenser Caps for Blood Collection Tubes, 13mm, in Bags
Catalog No. NC9873637
Encompass_Preferred
Dispenser; Blood Collection Tube; Tip Top™; Labcon; Dispenser cap with integral filter for 13mm blood collection tubes; Dispenses drops of filtered blood for clinical lab uses; Fits standard glass or plastic 13mm blood tubes; Polyethylene USP Classs VI resin; 1000 per bag/5 bags per case; Made in USA
Catalog No. NC9873637 Supplier Labcon North America Supplier No. 55118000009
Dispenser; Blood Collection Tube; Tip Top™; Labcon; Dispenser cap with integral filter for 13mm blood collection tubes; Dispenses drops of filtered blood for clinical lab uses; Fits standard glass or plastic 13mm blood tubes; Polyethylene USP Classs VI resin; 1000 per bag/5 bags per case; Made in USA