Learn More
YUKON MEDICAL 13mm Vialok Vented (100 per case) RX
SDP
Catalog No. NC1649456
Encompass_Preferred
13mm Vialok Vented Vial Access Devices feature a 0.2 micron filter that neutralizes vial pressure and minimizes aerosols and surface contamination. In addition to their superior vial security, they utilize a dual lumen spike, making for efficient transfers and minimizing the drug residual left in the vial.
This item is not returnable. View return policy
Catalog No. NC1649456 Supplier YUKON MEDICAL Supplier No. YM011
13mm Vialok Vented Vial Access Devices feature a 0.2 micron filter that neutralizes vial pressure and minimizes aerosols and surface contamination. In addition to their superior vial security, they utilize a dual lumen spike, making for efficient transfers and minimizing the drug residual left in the vial.