Learn More
Cayman Chemical 13-methyl Myristic Acid methyl ester, Purity: ≥98%, CAS: 5129-59-9, 5 mg
SDP
Catalog No. 503015864
Encompass_Preferred
A methylated bacterial fatty acid methyl ester; has been used as a marker of bacteria in sewage sludges and of bacterial contaminants in pharmaceutical manufacturing by GC-MS
Catalog No. 50-301-5864 Supplier Cayman Chemical Supplier No. 248125 MG
A methylated bacterial fatty acid methyl ester; has been used as a marker of bacteria in sewage sludges and of bacterial contaminants in pharmaceutical manufacturing by GC-MS