Learn More
Abnova™ 13q12 CISH Probe
Catalog No. 89335562 Shop All Abnova Corporation Products
Change view
Click to view available options
Product Type:
13q12 CISH Probe
Catalog No. Product Type
89-335-562 13q12 CISH Probe
1 options
Catalog No. 89-335-562 Supplier Abnova™ Supplier No. CG0015
Add to Cart
Add to Cart
13q12 CISH Probe is designed for the qualitative detection of human chromosome 13 specific sequences in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH).
Specifications
Content And Storage The product is ready-to-use. No reconstitution, mixing, or dilution is required. Bring probe to room temperature (18 to 25°C) and mix briefly before use.
Description The probe is intended to be used in combination with the CISH Implementation Kit 1, which provides necessary reagents for specimen pretreatment and post-hybridization processing.
Product Type 13q12 CISH Probe
Includes Reagent Provided:
1. Digoxigenin-labeled polynucleotides targeting sequences mapping in 13q12.11* (chr13:20,609,044-20,776,358)
2. Formamide based hybridization buffer
*according to Human Genome Assembly GRCh37/hg19