Learn More
Enzo Life Sciences 13(S)-Hydroxyoctadeca-9Z,11E-dienoic acid (50µg). CAS: 29623-28-7
SDP
Supplier:  Enzo Life Sciences BMLHO0130050
Encompass_Preferred
Biosynthesis in rabbit peritoneal serous membranes, atherosclerotic plaques, healthy blood vessels, psoriatic skin, endothelial cells. Stimulates prostacyclin production by endothelial cells. Inhibits thromboxane production in human platelets. Inhibits platelet and tumor cell adhesion to endothelium. Antagonizes 12-HETE-induced tumor cell adhesion to endothelium. Stimulates DNA synthesis in BALB/c 3T3 fibroblasts. Alternative name: 13(S)-HODE. Formulation: Solution in ethanol. Purity: ≥99% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-1029