Learn More
Enzo Life Sciences 13(S)-Hydroperoxyoctadeca-9Z,11E-dienoic acid (1mg). CAS: 33964-75-9
SDP
Catalog No. 502011034
Encompass_Preferred
Suppresses activation of platelets. Induces endothelial injury. Constricts isolated coronary arteries. Inhibits prostacyclin synthase. Alternative name: 13(S)-HPODE. Formulation: Oil dissolved in ethanol. Purity: ≥98%. Long Term Storage: -80°C.
Catalog No. 50-201-1034 Supplier Enzo Life Sciences Supplier No. BMLHP0131000
Suppresses activation of platelets. Induces endothelial injury. Constricts isolated coronary arteries. Inhibits prostacyclin synthase. Alternative name: 13(S)-HPODE. Formulation: Oil dissolved in ethanol. Purity: ≥98%. Long Term Storage: -80°C.