Learn More
Sarstedt Inc 13X100 TUBE 8ML RB SPK 500/PK
Catalog No. NC1532996 Shop All Sarstedt Inc. Products
Encompass_Preferred
Tube, 8 ml, 100 x 13 mm, round base, PS, 500 pcs./StackPack
This item is not returnable. View return policy
Catalog No. NC1532996 Supplier Sarstedt Inc Supplier No. 55.467.300
Tube, 8 ml, 100 x 13 mm, round base, PS, 500 pcs./StackPack