Learn More
Bellco Glass, Inc. 13x100mm Test Tubes W/SC
SDP
Catalog No. 502330073
Encompass_Preferred
Sterile Screw Cap Test Tubes are ideal for cell culturing and other applications that require a sterile surface.Gamma radiation sterilized and shipped ready-to-useScrewcap has built-in sealing rings to provide leakproof closuresPolystyrene tubes provide clarity and allows the user to easily observe contentsPolystyrene tubes are certified to 8,200 × gItems are not available for shipment to Mexico. Diameter (mm)-13, Height (mm)-100, Capacity-8mL, Tube Material-Polystyrene (PS), Tube Color-Natural, Cap Material-Polyethylene (PE), Cap Color-Natural, Shape-Round bottom, Package Type-25/Bag, Carton Qty-500
Catalog No. 50-233-0073 Supplier Bellco Glass, Inc. Supplier No. 130000293
Sterile Screw Cap Test Tubes are ideal for cell culturing and other applications that require a sterile surface.Gamma radiation sterilized and shipped ready-to-useScrewcap has built-in sealing rings to provide leakproof closuresPolystyrene tubes provide clarity and allows the user to easily observe contentsPolystyrene tubes are certified to 8,200 × gItems are not available for shipment to Mexico. Diameter (mm)-13, Height (mm)-100, Capacity-8mL, Tube Material-Polystyrene (PS), Tube Color-Natural, Cap Material-Polyethylene (PE), Cap Color-Natural, Shape-Round bottom, Package Type-25/Bag, Carton Qty-500