Learn More
Andwin Scientific 13X18 SELF SEAL POUCH 100/PK
SDP
Catalog No. NC9811298
Encompass_Preferred
The 13 x 18" Convertors sterilization pouch are ideal for fast and easy presentation of sterile items. Each pouch has a chemical process indicator that inform the user of the sterilization process used. The pouches can be taped shut. This is sold in a pack of 800.
This item is not returnable. View return policy
Catalog No. NC9811298 Supplier Andwin Scientific Supplier No. 11213236
The 13 x 18" Convertors sterilization pouch are ideal for fast and easy presentation of sterile items. Each pouch has a chemical process indicator that inform the user of the sterilization process used. The pouches can be taped shut. This is sold in a pack of 800.