Learn More
Gawet Marble and Granite 13X20X2 MARBLE SLAB
Catalog No. NC9879351
Encompass_Preferred
VERMONT MARBLE SLAB, Dimensions: 13 in. x 20 in. x 2 in. thick, Weight: 70 lbs,The Vermont Marble Slab is for use on laboratory furniture, It SUBSTANTIALLY REDUCES VIBRATION AND SHOCK where a Danby Vermont Marble Balance
Catalog No. NC9879351 Supplier Gawet Marble and Granite Supplier No. 13X20X2
VERMONT MARBLE SLAB, Dimensions: 13 in. x 20 in. x 2 in. thick, Weight: 70 lbs,The Vermont Marble Slab is for use on laboratory furniture, It SUBSTANTIALLY REDUCES VIBRATION AND SHOCK where a Danby Vermont Marble Balance