Learn More
Sarstedt Inc Reagent and Centrifuge Tube, 13x75mm, 5mL, Polypropylene, 500/Pack
Catalog No. NC9217339 Shop All Sarstedt Inc. Products
Encompass_Preferred
Reagent and Centrifuge Tube, Volume: 5mL, Diameter: 13mm, Length: 75mm, Base shape: Round base, Type of closure: Push cap, Material: Polypropylene (PP), Colour: Transparent, Non-sterile tube, Autoclavable to 121degreeC
Catalog No. NC9217339 Supplier Sarstedt Inc Supplier No. 55.525
Reagent and Centrifuge Tube, Volume: 5mL, Diameter: 13mm, Length: 75mm, Base shape: Round base, Type of closure: Push cap, Material: Polypropylene (PP), Colour: Transparent, Non-sterile tube, Autoclavable to 121degreeC