Learn More
Cayman Chemical 13Z,16Z-Docosadienoic Acid, Purity: ≥98%, CAS: 17735-98-7, 10 mg
SDP
Catalog No. 502980731
Encompass_Preferred
A natural ω-6 PUFA; an agonist of FFAR4 (GPR120) that strongly inhibits the secretion of ghrelin by isolated mouse gastric cells
Catalog No. 50-298-0731 Supplier Cayman Chemical Supplier No. 2074910 MG
A natural ω-6 PUFA; an agonist of FFAR4 (GPR120) that strongly inhibits the secretion of ghrelin by isolated mouse gastric cells