Learn More
Avanti Polar Lipids 1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] (ammonium salt), 474922-82-2, MFCD02262246, 200mg
Catalog No. NC1331950
Encompass
Formula Weight: 2693.285, Purity >99%, Storage Temperature: -20°C.
Catalog No. NC1331950 Supplier Avanti Polar Lipids Supplier No. 880150P200MG
Formula Weight: 2693.285, Purity >99%, Storage Temperature: -20°C.