Learn More
Avanti Polar Lipids 14 :0PC (DMPC), 18194-24-6, 1g
Catalog No. NC0627226
Encompass
Molecular Formula C36H72NO8P, Purity >99%, Formula Weight 677.950, 1,2-dimyristoyl-sn-glycero-3-phosphocholine
Catalog No. NC0627226 Supplier Avanti Polar Lipids Supplier No. 850345P1G
Molecular Formula C36H72NO8P, Purity >99%, Formula Weight 677.950, 1,2-dimyristoyl-sn-glycero-3-phosphocholine