Learn More
Avanti Polar Lipids 14 :0PG 1 GRAM
Catalog No. NC0841304
Encompass
1,2-dimyristoyl-sn-glycero-3-phospho-(1'-rac-glycerol) (sodium salt). 200880-40-6
Catalog No. NC0841304 Supplier Avanti Polar Lipids Supplier No. 840445P1G
1,2-dimyristoyl-sn-glycero-3-phospho-(1'-rac-glycerol) (sodium salt). 200880-40-6