Learn More
Ace Glass, Inc. 14/10 100mm distillation column
SDP
Catalog No. 50979990
Encompass_Preferred
14/10 100mm long Vigreux distillation column. Length in millimeters refers to the effective length as measured from the lowest to the highest indent of the column.
Catalog No. 50-979-990 Supplier Ace Glass, Inc. Supplier No. 934535
14/10 100mm long Vigreux distillation column. Length in millimeters refers to the effective length as measured from the lowest to the highest indent of the column.